Learn More
BINDER™ Analog Output
Non-adjustable analog output
£663.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
30238335
|
BINDER™
8012-2371 |
KB 65, KB 130, KB 260, KB 470, KB 720, KB 1060 |
£663.00
Each |
Estimated Shipment: 24-03-2025
Log in to see stock. |
||||||
30238324
|
BINDER™
8012-2375 |
KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, KB PRO 1600 |
£663.00
Each |
Estimated Shipment: 24-03-2025
Log in to see stock. |
||||||
30238254
|
BINDER™
8012-2573 |
KB PRO 130, KB PRO 260, KB PRO 470, KB PRO 260, KB PRO 470 |
£663.00
Each |
Estimated Shipment: 24-03-2025
Log in to see stock. |
||||||
30238280
|
BINDER™
8012-2574 |
KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600 |
£663.00
Each |
Estimated Shipment: 24-03-2025
Log in to see stock. |
||||||
Description
- Analog output
- Not adjustable
- For temperature values or temperature and humidity values
- 4 to 20 mA
Specifications
Copper | |
Analog Output |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BINDER™ Analog Output