Learn More
BINDER™ Analog Output 4-20 mA, Temperature
Analog Output for temperature values. The output is not adjustable.
Brand: BINDER™ 8012-2188
Additional Details : Weight : 0.01100kg
Description
- Accessory for Series FP Avantgarde.Line Drying and Heating Chambers
Specifications
Series FP Avantgarde.Line Drying and Heating Chambers | |
Temperature | |
4 to 20 mA |
Analog Output | |
230/240 V |