Learn More
Electrothermal™ Analogue Power Controller
Control the power supply to one piece of laboratory equipment at a time. Analogue Power Controller is designed to control the heating of resistive loads for bench top operation.
Brand:  Electrothermal™ 36223-02
Product Code. 10724216
  • £295.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Each heating controller has 2 neon indicators: ''Power On'' white neon light and ''Mantle/Bunsen Heater On'' amber neon light.
  • Regulator control knob can be turned clockwise to increase power.
  • As the knob is turned, the heating controller's amber neon light will pulsate to show that power is being supplied to the equipment being controlled, e.g. mantles, heating tape.
  • The pulse frequency decreases as the regulator control knob setting is increased
  • At maximum setting, the amber neon is continually illuminated.
  • Includes a support rod clamp at the rear of each heating controller to take a standard 12.5 mm (1/2in.) diameter rod.
  • Short mains lead with an IEC socket to connect to the resistive load.
  • An accessory extension mains lead is available where remote operation is required, e.g. in a fume extraction unit.
  • Mountable on standard 12 mm diameter scaffold or retort stands, and can also be wall mounted using a mounting bracket or stand directly on the bench.
Specifications
Specifications
Analogue
130 x 95 x 105 mm
1800 W
Temperature Controller
Heating Mantles
230 V
0.42 kg
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Electrothermal™ Analogue Power Controller > Analogue Power Controller