Learn More
Analyte Mix, High Level, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep 606-XH
Code : NEW
Additional Details : Weight : 0.00100kg Transport : UN number : 1262 Chem class : 3 Pack group : II
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Bis(2-Ethylhexyl)phthalate | 117-81-7 | 2000μg/mL |
Butylbenzyl phthalate | 85-68-7 | 2000μg/mL |
Diethyl phthalate | 84-66-2 | 2000μg/mL |
Dimethyl phthalate | 131-11-3 | 2000μg/mL |
Di-n-butyl phthalate | 84-74-2 | 2000μg/mL |
Di-n-octyl phthalate | 117-84-0 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Analytes Mix (High Level) | |
2mL amber prescored glass ampule | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
For USEPA Method 606, 8060, 8061 |
USEPA Method 606, SW-846 Method 8060 | |
1 mL | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
Isooctane |
Safety and Handling
- Analytes Mix (High Level)
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 2,2,4-trimethylpentane, bis(2-ethylhexyl)phthalate, BBP, dibutyl phthalate, di-n-octyl phthalate
Product intended for laboratory use only