Learn More
Analytes, High Level, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep 5041-AH
Code : NEW
Additional Details : Weight : 0.05000kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
1,2,3-Trichloropropane | 96-18-4 | 2000μg/mL |
1,2-Dibromo-3-chloropropane | 96-12-8 | 2000μg/mL |
1,2-Dibromoethane | 106-93-4 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Analytes (High Level) | |
2mL amber prescored glass ampule | |
Methanol-P&T | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
USEPA Method 504.1 | |
For US EPA Method 504, 504.1 | |
1 mL | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
Safety and Handling
- Regular Unleaded Gasoline (High Level)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H340-May cause genetic defects.
- H351-Suspected of causing cancer.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : gasoline, methanol
Product intended for laboratory use only