missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, 0.01M, Analytical Volumetric Solution, Reagecon™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
1 L
Packaging:
1L
Description
Ready to use aqueous volumetric solution for all analytical laboratory applications, which can be used for manual or automatic titrations. This product is certified to a specification as outlined in the Certificate of Analysis. Lot Number and Expiry Date are printed on the label. Solution is certified traceable to N.I.S.T and this will be stated in the Certificate of Analysis.
- Molecular Formula: Na2S2O3 • 5H2O
- Molar Mass: 248.18g/mol
- CAS: 10102-17-7
- EIN ECS No.: 231-867-5
Specifications
Boiling Point | 100°C |
Melting Point | 48.3°C |
CAS | 10102-17-7 |
Color | Colorless |
Physical Form | Solution |
Quantity | 1 L |
Molecular Formula | H10Na2O8S2 |
Synonym | sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate |
Solubility Information | Fully miscible |
InChI Key | PODWXQQNRWNDGD-UHFFFAOYSA-L |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'