Learn More
Abeba™ Anatom 32120 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand: Abeba™ 32120/43
82.60 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Additional Details : Weight : 1.44300kg
Description
- CE EN ISO 20345:2011 SB A, E SRC
- Microfiber
- White
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3582)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Breathable lining with Silver ions
- Instep area adjustable, with elastic band
- velcro fastening
- suitable for heel and foot lift
- Suitable for hygienically demanding areas
Specifications
CE EN ISO 20345:2011 SB A, E SRC | |
Microfiber | |
Slip-resistant | |
32120 |
White | |
43 | |
Polyurethane | |
Steel |