Learn More
Abeba™ Anatom 32130 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
£93.87 - £130.59
Specifications
Certifications/Compliance | CE EN ISO 20345:2011 S2 SRC |
---|---|
Color | White |
Material | Microfiber |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15280925
|
Abeba™
32130/36 |
36 |
£93.87
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15630355
|
Abeba™
32130/37 |
37 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15200935
|
Abeba™
32130/38 |
38 |
£93.87
1 pair |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
15650355
|
Abeba™
32130/39 |
39 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15660355
|
Abeba™
32130/40 |
40 |
£96.70
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15230935
|
Abeba™
32130/41 |
41 |
£93.87
1 pair |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
15680355
|
Abeba™
32130/42 |
42 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15690355
|
Abeba™
32130/43 |
43 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15260935
|
Abeba™
32130/44 |
44 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15610365
|
Abeba™
32130/45 |
45 |
£93.87
1 pair |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
15620365
|
Abeba™
32130/46 |
46 |
£93.87
1 pair |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
15290935
|
Abeba™
32130/47 |
47 |
£93.87
1 pair |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
15640365
|
Abeba™
32130/48 |
48 |
£93.87
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15210945
|
Abeba™
32130/49 |
49 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15220945
|
Abeba™
32130/50 |
50 |
£93.87
1 pair |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
15607678
|
Abeba™
32130/51 |
51 |
£93.87
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15617678
|
Abeba™
32130/52 |
52 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15200945
|
Abeba™
32130/48 |
48 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- CE EN ISO 20345:2011 S2 SRC
- Microfiber
- White
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area adjustable, with elastic band
- velcro fastening
- Suitable for heel and foot lift
Specifications
CE EN ISO 20345:2011 S2 SRC | |
Microfiber | |
Polyurethane | |
Steel |
White | |
Slip-resistant | |
32130 |