Learn More
Abeba™ Anatom 32135 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
£93.87
Specifications
Certifications/Compliance | CE EN ISO 20345:2011 S2 SRC |
---|---|
Color | Black |
Material | Microfiber |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15650365
|
Abeba™
32135/36 |
36 |
£93.87
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15660365
|
Abeba™
32135/37 |
37 |
£93.87
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15670365
|
Abeba™
32135/38 |
38 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15680365
|
Abeba™
32135/39 |
39 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15690365
|
Abeba™
32135/40 |
40 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15600375
|
Abeba™
32135/41 |
41 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15610375
|
Abeba™
32135/42 |
42 |
£93.87
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15620375
|
Abeba™
32135/43 |
43 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15630375
|
Abeba™
32135/44 |
44 |
£93.87
1 pair |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
15640375
|
Abeba™
32135/45 |
45 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15650375
|
Abeba™
32135/46 |
46 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15660375
|
Abeba™
32135/47 |
47 |
£93.87
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15670375
|
Abeba™
32135/48 |
48 |
£93.87
1 pair |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15862425
|
Abeba™
32135/49 |
49 |
£93.87
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15627678
|
Abeba™
32135/50 |
50 |
£93.87
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15637678
|
Abeba™
32135/51 |
51 |
£93.87
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
15647678
|
Abeba™
32135/52 |
52 |
£93.87
1 pair |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
Description
- CE EN ISO 20345:2011 S2 SRC
- Microfiber black
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area adjustable, with elastic band
- velcro fastening
- Suitable for heel and foot lift
Specifications
CE EN ISO 20345:2011 S2 SRC | |
Microfiber | |
Polyurethane | |
Steel |
Black | |
Slip-resistant | |
32135 |