Learn More
IKA™ Anchor stirrer
Stainless steel construction. For use with Lab reactor
Brand: IKA™ 0004663000
Additional Details : Weight : 0.12140kg
Description
- Minimum usable volume when homogenizing: 400 mL
- Stainless steel
- For use with Lab reactors
Specifications
Laboratory Reactor | |
LR 1000.11 | |
Anchor | |
Overhead Stirrer Shaft |