Learn More
Lenz Laborglasintrumente Anchor Stirrers With Journal
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 06321073
Additional Details : Weight : 0.25000kg
Description
- Made of precision calibrated glass tube
- Ground and polished bearing surface
- For flange vessels
- Made of DURAN™ tubing
Specifications
Lab Flange 100 LF | |
630 mm | |
Duran Glass |