Learn More
Lenz Laborglasintrumente Anchor Stirrers With Journal
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 06321579
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11506884
  • £148.00 / Each
Estimated Shipment: 14-08-2024
to see stock.
Add to basket
Description
Description
  • Made of precision calibrated glass tube
  • Ground and polished bearing surface
  • For flange vessels
  • Made of DURAN™ tubing
Specifications
Specifications
10 mm
Lab Flange 150 LF
700 mm
Duran Glass