missing translation for 'onlineSavingsMsg'
Learn More

Androsterone, 97%, Thermo Scientific Chemicals

Product Code. 10601545
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-41-8
Molecular Formula C19H30O2
Molecular Weight (g/mol) 290.447
InChI Key QGXBDMJGAMFCBF-HLUDHZFRSA-N
Synonym androsterone, androkinine, androtine, 5alpha-androsterone, 3alpha-hydroxy-5alpha-androstan-17-one, atromide ici, 3-epihydroxyetioallocholan-17-one, 3alpha-hydroxyetioallocholan-17-one, 5alpha-androstane-3alpha-ol-17-one, cis-androsterone
PubChem CID 5879
ChEBI CHEBI:16032
IUPAC Name (3R,5S,8R,9S,10S,13S,14S)-3-hydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-one
SMILES CC12CCC(CC1CCC3C2CCC4(C3CCC4=O)C)O

Specifications

Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 633
Specific Rotation Condition + 96.00 (20.00°C c=1,C2H5OH)
Specific Rotation +90° to +98° (20°C, 589nm) (c=0.7, ethanol)
Formula Weight 290.44
Percent Purity ≥96.0%
Physical Form Crystals or Powder
Chemical Name or Material Androsterone, 97%
Show More Show Less
Product Identifier
  • Androsterone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.