missing translation for 'onlineSavingsMsg'
Learn More

Andwin Scientific Lazy L-shaped Spreader

Product Code. 11765249 Shop All Cole-Parmer Products
Click to view available options
Color:
Blue
Unit Size:
Pack of 500
This item is not returnable. View return policy

Product Code. 11765249

Brand: Andwin Scientific 1420100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

L–shaped for a smooth and even sample distribution.

  • Polystyrene spreader
  • Durable disposable and non–breakable
  • Shape reduces the likelihood of the spreader digging into the agar
  • Pre–sterilized
  • Blue

Specifications

Disposable Single-use
Autoclavable Not Autoclavable
Color Blue
Shape L-Shape
Material Polystyrene
Product Type Cell Spreader
Sterility Sterile
For Use With (Application) Allows for a smooth and even sample distribution, reducing the likelihood of the spreader digging into the agar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.