missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Angled Three-Neck Round-Bottom Flasks

Three-neck distillation flasks with two angled side necks (20°)

Brand:  Lenz Laborglasintrumente™ 03315488

 View more versions of this product

Product Code. 10458402

  • £501.00 / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Acc. to DIN 12394 and in addition to DIN standard
  • With two angled side necks (20°)
  • With conical ground joints
  • With inscription label
  • Material: DURAN™
Specifications

Specifications

Center neck- NS 45/40, Side necks - NS 29/32
Borosilicate Glass (3.3)
1
279 mm
10,000 mL
Three-necks with two angled side necks
Distillation Flask
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.