missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANGPTL7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA536575
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 40 kDa. Purity is >95% by SDS-PAGE.
ANGPTL7 Polyclonal antibody specifically detects ANGPTL7 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
ANGPTL7 | |
Polyclonal | |
Unconjugated | |
ANGPTL7 | |
angiopoietin like 7; angiopoietin-like 7; Angiopoietin-like factor; angiopoietin-like factor (CDT6); angiopoietin-like protein 7; Angiopoietin-related protein 7; ANGPTL7; AngX; CDT6; Cornea-derived transcript 6 protein; dJ647M16.1; UNQ313/PRO356 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10218, 654812 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O43827, Q8R1Q3 | |
ANGPTL7 | |
Synthetic peptide corresponding to amino acids 321-370 of human Angptl7 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |