Learn More
Velp Scientifica™ Anhydrone
Velp Scientifica™ Anhydrone for use with Dumas Nitrogen Analyzer NDA 701 and 702.
Brand: Velp Scientifica™
Code : NEW
Additional Details : Weight : 0.45400kg Transport : UN number : 1475 Chem class : 5.1 Pack group : II
Description
- For use with NDA 701 and 702
Specifications
Anhydrone | |
454 g | |
Dumas Nitrogen Analyzer | |
0.454 kg |
Safety and Handling
- Magnesium perchlorate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P220-Keep/Store away from clothing/combustible materials.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : magnesium perchlorate