missing translation for 'onlineSavingsMsg'
Learn More

Aniline-2,5-disulfonic acid, 95%, Thermo Scientific Chemicals

Product Code. 11487026
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Aniline-2,5-disulfonic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aniline-2,5-disulfonic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-44-2
Molecular Formula C6H7NO6S2
Molecular Weight (g/mol) 253.243
MDL Number MFCD00035753
InChI Key LDCCBULMAFILCT-UHFFFAOYSA-N
Synonym 2-amino-1,4-benzenedisulfonic acid, aniline-2,5-disulfonic acid, 2,5-disulfoaniline, 4-sulfometanilic acid, 1,4-benzenedisulfonic acid, 2-amino, 2,5-disulfo-1-aminobenzene, 2-amino-1,4-disulfobenzene, 1-anilino-2,5-disulfonic acid, 2-amino-p-benzenedisulfonic acid, unii-5e2ot6t2ps
PubChem CID 60988
IUPAC Name 2-aminobenzene-1,4-disulfonic acid
SMILES C1=CC(=C(C=C1S(=O)(=O)O)N)S(=O)(=O)O

Specifications

Melting Point >300°C
Quantity 5 g
Formula Weight 253.25
Percent Purity 95%
Chemical Name or Material Aniline-2,5-disulfonic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'