missing translation for 'onlineSavingsMsg'
Learn More

Aniline-2-sulfonic Acid, 95%, Thermo Scientific Chemicals

Product Code. 10147180
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-21-1
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.19
MDL Number MFCD00007705
InChI Key ZMCHBSMFKQYNKA-UHFFFAOYSA-N
Synonym orthanilic acid, aniline-2-sulfonic acid, o-aminobenzenesulfonic acid, o-sulfanilic acid, aniline-o-sulfonic acid, 2-sulfanilic acid, aniline-o-sulphonic acid, 1-aminobenzene-2-sulfonic acid, 2-aminobenzenesulphonic acid, benzenesulfonic acid, 2-amino
PubChem CID 6926
ChEBI CHEBI:1015
IUPAC Name 2-aminobenzenesulfonic acid
SMILES C1=CC=C(C(=C1)N)S(=O)(=O)O

Specifications

Melting Point >300°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula H2NC6H4SO3H
Quantity 25 g
Beilstein 14, 681
Merck Index 15, 6978
Solubility Information Solubility in water: 3100mg/l (23°C)
Formula Weight 173.19
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Aniline-2-sulfonic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.