missing translation for 'onlineSavingsMsg'
Learn More

Aniline, 99+%

Product Code. p-7021911
Click to view available options
Quantity:
100 g
1000 g
5000 g
Unit Size:
1000g
100g
5000g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 15483087

Brand: Thermo Scientific Alfa Aesar A14443.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 62-53-3 | C6H7N | 93.13 g/mol

Aniline is used as a solvent. It has been used as an antiknock compound for gasolines. It has a major application in the preparation of polyurethane which is used in making plastic, building thermal foam insulations and in refrigerators. It is widely used in the preparation of methylene dianiline and related compounds by reacting with formaldehyde. It combines with nitric acid and is utilized as rocket fuel for small missiles and jet assisted take-off. It acts as an additive to rubber and its derivatives such as phenylenediamines and diphenylamine are used as antioxidants. It finds application in dyes and pigments in agricultural and polymer industries. It is employed at a smaller scale in the production of the intrinsically conducting polymer polyaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aniline is used as a solvent. It has been used as an antiknock compound for gasolines. It has a major application in the preparation of polyurethane which is used in making plastic, building thermal foam insulations and in refrigerators. It is widely used in the preparation of methylene dianiline and related compounds by reacting with formaldehyde. It combines with nitric acid and is utilized as rocket fuel for small missiles and jet assisted take-off. It acts as an additive to rubber and its derivatives such as phenylenediamines and diphenylamine are used as antioxidants. It finds application in dyes and pigments in agricultural and polymer industries. It is employed at a smaller scale in the production of the intrinsically conducting polymer polyaniline.

Solubility
Miscible with water.

Notes
Light sensitive. Incompatible with oxidizing agents, iron and iron salts.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 62-53-3
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00007629
InChI Key PAYRUJLWNCNPSJ-UHFFFAOYSA-N
Synonym benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
PubChem CID 6115
ChEBI CHEBI:17296
IUPAC Name aniline
SMILES NC1=CC=CC=C1

Specifikationer

Melting Point -6°C
Density 1.021
Boiling Point 183°C to 184°C
Flash Point 70°C (158°F)
Refractive Index 1.586
Quantity 100 g
UN Number UN1547
Beilstein 605631
Merck Index 14,659
Solubility Information Miscible with water.
Formula Weight 93.13
Percent Purity ≥99%
Chemical Name or Material Aniline
Visa mer Visa mindre
compliance-icons compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • Aniline
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Faroangivelser
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Kompletterande information
  • MIXTURE LIST-Contains : anilin

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.