missing translation for 'onlineSavingsMsg'
Learn More
Aniline, 99.8%, pure, Thermo Scientific Chemicals Product Code.: 10132970

Aniline, 99.8%, pure, Thermo Scientific Chemicals

Product Code. 10132970
5 mL, Glass bottle
Click to view available options
Quantity:
5 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10132970

Brand: Thermo Scientific Acros 158190050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-53-3
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00007629
InChI Key PAYRUJLWNCNPSJ-UHFFFAOYSA-N
Synonym benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
PubChem CID 6115
ChEBI CHEBI:17296
IUPAC Name aniline
SMILES NC1=CC=CC=C1

Specifications

Melting Point -6.2°C
Color Pink to White
Density 1.0213g/mL
Boiling Point 181.0°C to 185.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 99.75% min. (GC)
Packaging Glass bottle
Linear Formula C6H5NH2
Refractive Index 1.5845 to 1.5865
Quantity 5 mL
Beilstein 12, 59
Fieser 06,21; 17,16
Merck Index 15, 652
Specific Gravity 1.0213
Solubility Information Solubility in water: 36g/L (20°C).
Viscosity 4.4 mPa.s (20°C)
Formula Weight 93.13
Vapor Pressure 0.5mmHg at 20°C
Percent Purity 99.8%
Grade Pure
Physical Form Crystalline Powder, Crystals or Crystalline Mass
Chemical Name or Material Aniline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465039 Certificate TypeCertificate of Analysis Date31/07/2024 Catalog Number158190010,158190025,158191000,158190050
Lot NumberA0462447 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog Number158190010,158190025,158191000,158190050
Lot NumberA0458742 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number158190010,158190025,158191000,158190050
Lot NumberA0458741 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number158190010,158190025,158191000,158190050
Lot NumberA0440437 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog Number158190010,158190025,158191000,158190050
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Aniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Aniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aniline, 99.8%, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.