missing translation for 'onlineSavingsMsg'
Learn More

Aniline hydrochloride, 99%

Product Code. 10001760 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10001760

Brand: Thermo Scientific Acros 158411000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-04-1 | C6H8ClN | 129.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-04-1
Molecular Formula C6H8ClN
Molecular Weight (g/mol) 129.59
MDL Number MFCD00012958
InChI Key MMCPOSDMTGQNKG-UHFFFAOYSA-N
Synonym aniline hydrochloride, aniline chloride, benzenamine, hydrochloride, anilinium chloride, aniline.hcl, phenylamine hydrochloride, chlorid anilinu, phenylammonium chloride, sul anilinova, chlorhydrate d'aniline
PubChem CID 8870
IUPAC Name aniline;hydrochloride
SMILES [H+].[Cl-].NC1=CC=CC=C1

Specifications

Melting Point 196.0°C to 199.0°C
Color Cream-Yellow to White
Boiling Point 245.0°C
Flash Point 193°C
Loss on Drying 0.5 % max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5NH2·HCl
Quantity 100 g
Merck Index 15,652
Solubility Information Solubility in water: 1070g/L (25°C). Other solubilities: insoluble in ethyl ether and chloroform
Formula Weight 129.59
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Aniline hydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.