Learn More
Aniline hydrochloride, 99%
Alle ansehen Thermo Scientific Chemicals ProdukteBeschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Melting Point | 196.0°C to 199.0°C |
| Color | Cream-Yellow to White |
| Boiling Point | 245.0°C |
| Flash Point | 193°C |
| Loss on Drying | 0.5 % max. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Plastic bottle |
| Linear Formula | C6H5NH2·HCl |
| Quantity | 2.5 kg |
| Mehr anzeigen |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
| Lot Number | Certificate Type | Date | Product Code |
|---|---|---|---|
| A0464346 | Certificate of Analysis | 04/07/2024 | 158415000,158411000,158410025 |
| A0443422 | Certificate of Analysis | 28/08/2023 | 158415000,158411000,158410025 |
| A0453135 | Certificate of Analysis | 16/05/2023 | 158415000,158411000,158410025 |
| A0374166 | Certificate of Analysis | 15/12/2022 | 158415000,158411000,158410025 |
| A0392136 | Certificate of Analysis | 15/12/2022 | 158415000,158411000,158410025 |
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.
