missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Animal-Free Human Hematopoietic Stem Cell Expansion Cytokine Package (IL-3), PeproTech®

The Animal-Free Human Hematopoietic Stem Cell Expansion Cytokine Package contains the key components required for ex vivo human hematopoietic stem cells.

Brand:  Gibco™ AF-HHSC3

1822.80 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 17941861

  • £1960.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Animal-Free Human Hematopoietic Stem Cell Expansion Cytokine Package contains the key components required for ex vivo human hematopoietic stem cells.

The package contains the following four vials:
• 100 μg Animal-Free Human Flt3 Ligand (Cat. No. AF-300-19-100UG)
• 100 μg Animal-Free Human SCF (Cat. No. AF-300-07-100UG)
• 100 μg Animal-Free Human TPO (Cat. No. AF-300-18-100UG)
• 10 μg Animal-Free Human IL-3 (Cat. No. AF-200-03-10UG)

Flt3 Ligand is a growth factor that regulates proliferation of early hematopoietic cells. Flt3 Ligand by itself does not stimulate proliferation of early hematopoietic cells, but synergizes with other CSFs and interleukins to induce growth and differentiation.

SCF is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. SCF and c-Kit are essential for the survival, proliferation, and differentiation of hematopoietic cells committed to the melanocyte and germ cell lineages. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells.

TPO is a lineage-specific growth factor produced in the liver, kidney, and skeletal muscle. It stimulates the proliferation and maturation of megakaryocytes and promotes increased circulating levels of platelets in vivo.

IL-3 is a hematopoietic growth factor that promotes the survival, differentiation, and proliferation of committed progenitor cells of the megakaryocyte, granulocyte-macrophage, erythroid, eosinophil, basophil, and mast cell lineages.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Functional Assay
Stem Cell Expansion Cytokine Package
Human
Lyophilized
1 pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number06152012 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAF-HHSC3
Lot Number06142012 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAF-HHSC3
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Animal-Free Human Hematopoietic Stem Cell Expansion Cytokine Package (IL-3), PeproTech® > 1 Pack

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.