Learn More
Gibco™ Animal-Free Human Hematopoietic Stem Cell Expansion Cytokine Package (IL-6), PeproTech®
The Animal-Free Human Hematopoietic Stem Cell Expansion Cytokine Package contains the key components required for ex vivo human hematopoietic stem cells.
Brand:  Gibco™ AF-HHSC6
1822.80 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Product Code. 17911551
  • £1960.00 / Each
Explore more special offers
Description
Description
The Animal-Free Human Hematopoietic Stem Cell Expansion Cytokine Package contains the key components required for ex vivo human hematopoietic stem cells.
The package contains the following four vials:
• 100 μg Animal-Free Human Flt3 Ligand (Cat. No. AF-300-19-100UG)
• 100 μg Animal-Free Human SCF (Cat. No. AF-300-07-100UG)
• 100 μg Animal-Free Human TPO (Cat. No. AF-300-18-100UG)
• 20 μg Animal-Free Human IL-6 (Cat. No. AF-200-06-20UG)
Flt3 Ligand is a growth factor that regulates proliferation of early hematopoietic cells. Flt3 Ligand by itself does not stimulate proliferation of early hematopoietic cells, but synergizes with other CSFs and interleukins to induce growth and differentiation.
SCF is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. SCF and c-Kit are essential for the survival, proliferation, and differentiation of hematopoietic cells committed to the melanocyte and germ cell lineages. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells.
TPO is a lineage-specific growth factor produced in the liver, kidney, and skeletal muscle. It stimulates the proliferation and maturation of megakaryocytes and promotes increased circulating levels of platelets in vivo.
IL-6 is a pleiotropic cytokine that stimulates B cell differentiation and antibody production, synergizes with IL-3 in megakaryocyte development and platelet production, induces expression of hepatic acute-phase proteins, and regulates bone metabolism.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Functional Assay
Stem Cell Expansion Cytokine Package
Human
Lyophilized
1 pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number06152012 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAF-HHSC6
Special Offers
Special Offers
Product Content Correction
Product Title
Gibco™ Animal-Free Human Hematopoietic Stem Cell Expansion Cytokine Package (IL-6), PeproTech® > 1 Pack