Learn More
Laboratoires Anios™ Aniosgel 85 NPC
Laboratoires Anios™ Aniosgel 85 NPC contains no allergenic substances, fragrance, or coloring, and is suitable for hygienic treatment and surgical disinfection of hands
Brand: Laboratoires Anios™ 1644333
Description
- No allergenic substances, fragrance, or coloring
- Broad antimicrobial spectrum
- Hygienic treatment in 30 sec.
- For hygienic treatment and surgical disinfection
- Active against bacteria, mycobacteria, yeasts, molds, viruses
Specifications
Sterilizer/Disinfectant Hand Gel | |
Ethanol in the presence of thickening, moisturizing and emollient agents, and water. | |
Airless bottles | |
1 L |
EN 13727, EN 13727, EN 14348, EN 13624, EN 14476, EN 14476, EN 1500, EN 12791 | |
Hygienic treatment and surgical disinfection of hands | |
12 Bottles |
Safety and Handling
- ANIOSGEL 85 NPC
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403-Store in a well-ventilated place.
Supplemental information
- MIXTURE LIST-Contains : ETHYL ALCOHOL, PROPAN-2-OL