Learn More
Laboratoires Anios Aniosgel 85 NPC; Eighty-Five NPC Disinfecting Gel
Thixotropic hydroalcoholic gel for hygienic treatment and surgical disinfection by hand-rubbing.
Brand: Laboratoires Anios 1644638UZ
Code : NEW
Additional Details : CAS Number : 64-17-5 Weight : 0.86000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Description
Disinfecting gel for hydroalcoholic hand-rubbing:
- Ethyl alcohol: 755 ml/l
- Flash point: +23°C
- Density at +20°C: approx. 0.862
- pH at +20°C: approx. 5.5 (near to skin pH)
- Viscosity at +20°C: from 1200 to 2500 mPa.s
- Storage: from +5°C and +25°C
- Aerobiocontamination study available on request
Specifications
Sanitizer | |
Hydroalcoholic thixotropic gel for hygienic treatment and surgical hand | |
1 L |
Safety and Handling
- ANIOSGEL 85 NPC
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403-Store in a well-ventilated place.
Supplemental information
- MIXTURE LIST-Contains : ETHYL ALCOHOL, PROPAN-2-OL