missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anisil, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Anisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Anisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Anisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | 132°C to 134°C |
Quantity | 5 g |
Beilstein | 531345 |
Solubility Information | Insoluble in water. |
Formula Weight | 270.29 |
Percent Purity | ≥98% |
Chemical Name or Material | Anisil |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'