missing translation for 'onlineSavingsMsg'
Learn More

Anisole, 99%, pure, Thermo Scientific Chemicals

£19.60 - £103.52

Chemical Identifiers

CAS 100-66-3
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
InChI Key RDOXTESZEPMUJZ-UHFFFAOYSA-N
Synonym methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
PubChem CID 7519
ChEBI CHEBI:16579
IUPAC Name anisole
SMILES COC1=CC=CC=C1
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10265720
View Documents Promotion Details
Thermo Scientific Acros
153920050
5 mL Glass bottle
£19.60
5mL
Please to purchase this item. Need a web account? Register with us today!
10458500
View Documents
Thermo Scientific Acros
153922500
250 mL Glass bottle
£20.90
250mL
Please to purchase this item. Need a web account? Register with us today!
10347710
View Documents
Thermo Scientific Acros
153920010
1 L Glass bottle
£49.25
1 L
Please to purchase this item. Need a web account? Register with us today!
11984711
View Documents
Thermo Scientific Acros
153920025
2.5 L Glass bottle
£103.52
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

100-66-3
108.14
methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
CHEBI:16579
COC1=CC=CC=C1
C7H8O
RDOXTESZEPMUJZ-UHFFFAOYSA-N
7519
anisole

Specifications

100-66-3
Colorless
154.0°C
Authentic
C7H8O
1.5160 to 1.5190
01,41
0.99
Solubility in water: 1.6g/L (20°C). Other solubilities: soluble in alcohol, ether, acetone
COC1=CC=CC=C1
108.14
CHEBI:16579
99%
Liquid
-37.0°C
0.9900g/mL
43°C
98.5% min. (GC)
C6H5OCH3
06,138
15,662
methoxybenzene, methyl phenyl ether, benzene, methoxy, anisol, phenyl methyl ether, phenoxymethane, anizol, phenol methyl ether, methoxy-benzene
RDOXTESZEPMUJZ-UHFFFAOYSA-N
anisole
7519
108.14
Pure
Anisole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ven

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 202-876-1

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.