missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anisole, Pure, Fisher Chemical
Click to view available options
500mL, Plastic Coated Glass bottle
Specifications
CAS | 100-66-3 |
Melting Point | -37°C |
Color | Colorless |
Boiling Point | 154°C |
Packaging | Plastic Coated Glass bottle |
Molecular Formula | C7H8O |
MDL Number | 8354 |
Quantity | 500 mL |
Vapor Density | 3.72 |
Formula Weight | 108.14g/mol |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
Product Identifier
- Anisole
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'