missing translation for 'onlineSavingsMsg'
Learn More
Anisomycin, Tocris Bioscience™ Product Code.: 18712664

Anisomycin, Tocris Bioscience™

Product Code. 18712664
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18712664

Brand: Tocris Bioscience 1290/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

JNK, SAPK and p38 activator

Anisomycin is a protein synthesis inhibitor (blocks translation). Potent activator of stress-activated protein kinases (JNK/SAPK) and p38 MAP kinase. Acts as a potent signaling agonist to selectively elicit homologous desensitization of immediate early gene induction (c-fos, fosB, c-jun, junB and junD).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22862-76-6
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00077650
InChI Key YKJYKKNCCRKFSL-BFHYXJOUSA-N
Synonym (2R,3S,4S)-2-[(4-Methoxyphenyl)methyl]-3,4-pyrrolidinediol 3-acetate
PubChem CID 253602
ChEBI CHEBI:338412
IUPAC Name (2S,3R,4R)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl acetate
SMILES COC1=CC=C(C[C@@H]2NC[C@@H](O)[C@@H]2OC(C)=O)C=C1

Specifications

Type Anisomycin
Quantity 10 mg
Formula Weight 265.31
Percent Purity >98%
Chemical Name or Material Anisomycin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Anisomycin, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.