Learn More
Emma Safety Footwear Anne Safety Shoes
Supremely comfortable, sculpted ladies safety shoe. Features SmartTecAir™ technology to remove moisture and perspiration instantly.
Brand: Emma Safety Footwear MM92251636
This item is not returnable.
View return policy
Description
- S1P-Safety shoe with sole plate for dry work
- GripForce™ Easy Twist
- Microfiber construction
- ESD: Electrostatic Discharge
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
36 |
Laces | |
Anne | |
Microfiber | |
Steel |