missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Annexin V Conjugates for Apoptosis Detection
Detect early stages of apoptosis with Annexin V stand-alone Alexa Fluor, APC, Pacific Blue, PE, FITC, and biotin conjugates using flow cytometry.
£365.00 - £744.00
Specifications
Product Type | Annexin V conjugate |
---|---|
Shipping Condition | Wet Ice |
For Use With (Equipment) | Flow Cytometer |
Product Code | Brand | Conjugate | Excitation/Emission | Flow Cytometer Laser Lines | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Conjugate | Excitation/Emission | Flow Cytometer Laser Lines | Price | Quantity & Availability | |||
10633853
|
Molecular Probes™
A13201 |
Alexa Fluor 488 | 495/519 | 488 |
£538.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10267242
|
Molecular Probes™
A13199 |
FITC | 494/518 | 488 |
£504.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10277242
|
Molecular Probes™
A13202 |
Alexa Fluor 568 | 578/603 | 532, 561 |
£528.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10443442
|
Molecular Probes™
A13203 |
Alexa Fluor 594 | 590/617 | 532 |
£530.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10318162
|
Molecular Probes™
A13204 |
Biotin-X |
£564.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10740194
|
Molecular Probes™
A23202 |
Alexa Fluor 350 | 346/442 | UV |
£530.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10297152
|
Molecular Probes™
A23204 |
Alexa Fluor 647 | 650/665 | 633-637 |
£522.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10267452
|
Molecular Probes™
A35108 |
Alexa Fluor 555 | 555/565 | 532, 561 |
£365.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10614263
|
Molecular Probes™
A35109 |
Alexa Fluor 680 | 679/702 | 633-637 |
£548.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10246572
|
Molecular Probes™
A35110 |
APC (Allophycocyanin) | 650/660 | 633-637 |
£744.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10033122
|
Molecular Probes™
A35111 |
PE | 565/578 | 488, 532, 561 |
£518.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10695373
|
Molecular Probes™
A35122 |
Pacific Blue | 410/455 | 405 |
£518.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Achieve quick and reliable detection of early cell apoptosis with Annexin V stand-alone conjugates for apoptosis detection. Annexin V conjugates offer up to 100-fold difference in fluorescence signal intensity between apoptotic and non-apoptotic cells using flow cytometry.
Annexin V has a high affinity for phosphatidylserine (PS), which becomes exposed on the outer leaflet of cells undergoing apoptosis. Because of this affinity, fluorescently labeled annexin V reagents are commonly used in apoptosis research.Annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, an indicator of intermediate stages of apoptosis. The difference in fluorescence intensity between apoptotic and nonapoptotic cells stained with our fluorescent annexin V conjugates, as measured by flow cytometry, is typically about 100-fold.
In collaboration with Nexins Research BV, we provide the best and brightest annexin V conjugates available, including Alexa Fluor 350, 488, 555, 568, 594, 647, and 680 annexin V conjugates, as well as Annexin V APC, Biotin-X, FITC, Pacific Blue, and PE conjugates. Highly fluorescent annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, one of the earliest indicators of apoptosis.
The Annexin V Pacific Blue conjugate is violet excitable, making it ideal for instruments with a violet laser and for multicolor experiments that include green- or red-fluorescent dyes.
The benefits of our annexin V conjugates include:
• Conjugated to Invitrogen Alexa Fluor and eFluor dyes for brighter signals
• Conjugates for all available lasers
• Available as stand-alone reagents or easy-to-use kits
Annexin V staining to detect apoptotic cells can only be done on live cells and tissue. If samples are to be fixed post-staining, there are specific conditions required to achieve transient retention of signal. These include use of an alcohol-free, aldehyde-based fixation method, use of buffers containing Ca2+ and avoidance of surfactants/detergents. For your convenience, we also offer a concentrated annexin-binding buffer that facilitates the binding of annexin V to phosphatidylserine in apoptosis assays.
Spécification
Annexin V conjugate | |
Wet Ice | |
Flow Cytometer |