missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Annexin V-FITC
Serves as a sensitive probe for flow cytometric analysis of cells that are undergoing apoptosis
£181.00 - £690.00
Specifications
Sample Volume | 5 μL |
---|---|
Storage Buffer | Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Content And Storage | 4°C |
For Use With (Application) | Flow Cytometry |
Conjugate | FITC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10186463
|
BD
556419 |
200 Tests |
£690.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15878688
|
BD
556420 |
100 Tests |
£426.00
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15850609
|
BD
560931 |
25 Tests |
£181.00
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Identifies apoptosis at an earlier stage than assays based on nuclear changes such as DNA fragmentation.
- Format: FITC
Flow Cytometry
Specifications
5 μL | |
4°C | |
FITC |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide | |
Flow Cytometry |
Research Use Only