missing translation for 'onlineSavingsMsg'
Learn More
Ansell AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves Product Code.: 11517853
Add to basket

Ansell AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves

Product Code. 11517853 Shop All Ansell Products
Size: 8; Quantity: 1/Pk
Click to view available options
Size: 8; Quantity: 1/Pk

Product Code. 11517853

Brand: Ansell 2100080

Add to basket
Add to basket

Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.

  • Provide flexibility and tactility
  • Made from an exceptionally thin chemical-resistant, five-layer laminated film
  • Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
  • Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
  • Hand specific
  • Broad spectrum chemical resistance
  • Wing-thumb for added wearer comfort
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 0
Certifications/Compliance (Europe) CE 0493, EN420:2003 + A1:2009; Category lll
Applications Industrial
Color White
Latex Free Yes
Material Laminated Film
Product Type Chemical-resistant Gloves
Size 8
Thickness (English) 2.5 mil
Thickness (Metric) 0.06 mm
EN388 Standard EN 388:0000
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ABCEFGHIMS
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Grip Design Smooth
Lining Unlined
Model 02-100_8
Series AlphaTec™ 02-100
Length (English) 15 in.
Length (Metric) 38 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.