missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves
Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Description
Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.
- Provide flexibility and tactility
- Made from an exceptionally thin chemical-resistant, five-layer laminated film
- Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
- Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
- Hand specific
- Broad spectrum chemical resistance
- Wing-thumb for added wearer comfort

Specifications
Specifications
Certifications/Compliance (Europe) | CE 0493, EN420:2003 + A1:2009; Category lll |
Material | Laminated Film |
Size | 7 |
Thickness (English) | 2.5 mil |
Thickness (Metric) | 0.06 mm |
EN388 Standard | EN 388:0000 |
EN ISO 374-1 | Type A |
EN ISO 374-1 CHEMICAL LETTER CODES | ABCEFGHIMS |
EN ISO 374 - 5 | Bacteria, Fungi WITH Virus Protection |
Grip Design | Smooth |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Documents
Scientific Resources
Brochures
Applications and solutions series: Safety edition
File size: 7.7 MB
Product Information
Datasheets
Ansell barrier glove 02 100 specification
File size: 200.6 kB
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves >
Spot an opportunity for improvement?Share a Content Correction