missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves

Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.

Brand:  Ansell 2100090

147.55 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 12333609

  • £208.00 / 12 pairs
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.

  • Provide flexibility and tactility
  • Made from an exceptionally thin chemical-resistant, five-layer laminated film
  • Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
  • Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
  • Hand specific
  • Broad spectrum chemical resistance
  • Wing-thumb for added wearer comfort

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE 0493, EN420:2003 + A1:2009; Category lll
Specific
Laminated Film
2.5 mil
EN 388:0000
ABCEFGHIMS
Gauntlet
Unlined
AlphaTec™ 02-100
38 cm
White
Yes
9
0.06 mm
Type A
Bacteria, Fungi WITH Virus Protection
Smooth
2-100 9 902212
15 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers