missing translation for 'onlineSavingsMsg'
Learn More
Ansell TouchNTuff™ DermaShield™ 73-701 Neoprene Cleanroom Gloves Product Code.: 11746713
Add to basket

Ansell TouchNTuff™ DermaShield™ 73-701 Neoprene Cleanroom Gloves
BIOPHARMA

Product Code. 11746713 Shop All Ansell Products
8
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
This item is not returnable. View return policy

Product Code. 11746713

Brand: Ansell 73701080

Add to basket
Add to basket
This item is not returnable. View return policy

Sterile neoprene (polychloroprene) cleanroom gloves are free of proteins and accelerators. Suitable for use in aseptic and Class 100/ISO 5 cleanroom environments.

  • Sterile disposable gloves feature a unique neoprene formulation for greater durability and puncture resistance.
  • Offers high levels of chemical splash resistance protecting from various hazards.
  • Beaded cuff designed with SUREFIT™ technology prevents cuffs from rolling down.
  • Latex-free and chemical accelerator-free eliminates Type I and Type IV allergy risks.
  • Recommended for transferring liquids and solids, sample taking, blending, compounding, and aseptic filling/labeling.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance 21 CFR 820 Quality System Regulations, ASTM D3577, ASTM D7160, ASTM D6978-05, ISO 10282, ISO 11193, ISO 13485, ISO 14001
Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009, EN 421:2010, EN 556-1:2001, EN ISO 374-1:2016, EN ISO 374-5:2016
Chemotherapeutic Rated Yes
Chlorinated Yes
Cleanroom Class Class 100 (ISO 5)
Accelerator Free Yes
AQL 0.65
Color Green
Disposable Single-use
Exterior Finish No Texture
Finger Thickness (English) 8.3 mil
Finger Thickness (Metric) 0.19 mm
Hand Specific
Latex Free Yes
Material Polychloroprene
Palm Thickness (English) 7 mil
Palm Thickness (Metric) 0.18 mm
Powdered Powder Free
Size 8
Sterility Sterile
Thickness (English) 8.3 mil
Thickness (Metric) 0.19 mm
Manufacturing/QMS Audit Standards ISO 9001:2015
Model 73-701
Series TouchNTuff™ 73-701
Tensile Strength 17 MPa
Length (English) 11.6 in.
Length (Metric) 29.5 cm
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2410078504 Certificate TypeCertificate of Irradiation Date17/04/2025 Catalog Number73701080
Lot Number2501033904 Certificate TypeCertificate of Irradiation Date26/03/2025 Catalog Number73701080
Lot Number2407004904 Certificate TypeCertificate of Irradiation Date14/01/2025 Catalog Number73701080
Lot Number2410078504 Certificate TypeCertificate of Compliance Date17/04/2025 Catalog Number73701080
Lot Number2501033904 Certificate TypeCertificate of Compliance Date26/03/2025 Catalog Number73701080
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell TouchNTuff™ DermaShield™ 73-701 Neoprene Cleanroom Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.