Learn More
Anthracene-9,10-diboronic acid bis(pinacol) ester, 95%, Thermo Scientific Chemicals
£56.60 - £210.00
Chemical Identifiers
CAS | 863992-56-7 |
---|---|
Molecular Formula | C26H32B2O4 |
Molecular Weight (g/mol) | 430.16 |
MDL Number | MFCD16294538 |
InChI Key | ZLXSWNVYGSZXOP-UHFFFAOYSA-N |
Synonym | 9,10-anthracenediboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracene, 4,4,5,5-tetramethyl-2-10-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracen-9-yl-1,3,2-dioxaborolane, amtb237, anthracene-9,10-diboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolanyl anthracene, 9,10-bis 4,4,5,5-tetramethyl 1.3.2 dioxaborolan-2-yl anthracene, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl anthracene, 1,3,2-dioxaborolane, 2,2'-9,10-anthracenediyl bis 4,4,5,5-tetramethyl |
PubChem CID | 57415691 |
IUPAC Name | 4,4,5,5-tetramethyl-2-[10-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracen-9-yl]-1,3,2-dioxaborolane |
SMILES | CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15459415
|
Thermo Scientific Alfa Aesar
H51921.03 |
1 g |
£56.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15469415
|
Thermo Scientific Alfa Aesar
H51921.06 |
5 g |
£210.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used synthesis, structure and electrochemistry of mono- and dinuclear osmium n,n-di- and tetraphenylbipyridyls and extended bipyridyls. It is also used as a primary and secondary intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used synthesis, structure and electrochemistry of mono- and dinuclear osmium n,n′-di- and tetraphenylbipyridyls and extended bipyridyls. It is also used as a primary and secondary intermediate.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
863992-56-7 | |
430.16 | |
ZLXSWNVYGSZXOP-UHFFFAOYSA-N | |
57415691 | |
CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CC=C12 |
C26H32B2O4 | |
MFCD16294538 | |
9,10-anthracenediboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracene, 4,4,5,5-tetramethyl-2-10-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracen-9-yl-1,3,2-dioxaborolane, amtb237, anthracene-9,10-diboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolanyl anthracene, 9,10-bis 4,4,5,5-tetramethyl 1.3.2 dioxaborolan-2-yl anthracene, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl anthracene, 1,3,2-dioxaborolane, 2,2'-9,10-anthracenediyl bis 4,4,5,5-tetramethyl | |
4,4,5,5-tetramethyl-2-[10-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracen-9-yl]-1,3,2-dioxaborolane |
Specifications
863992-56-7 | |
MFCD16294538 | |
ZLXSWNVYGSZXOP-UHFFFAOYSA-N | |
4,4,5,5-tetramethyl-2-[10-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)anthracen-9-yl]-1,3,2-dioxaborolane | |
57415691 | |
95% |
C26H32B2O4 | |
9,10-anthracenediboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracene, 4,4,5,5-tetramethyl-2-10-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl anthracen-9-yl-1,3,2-dioxaborolane, amtb237, anthracene-9,10-diboronic acid bis pinacol ester, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolanyl anthracene, 9,10-bis 4,4,5,5-tetramethyl 1.3.2 dioxaborolan-2-yl anthracene, 9,10-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl anthracene, 1,3,2-dioxaborolane, 2,2'-9,10-anthracenediyl bis 4,4,5,5-tetramethyl | |
CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=C(B2OC(C)(C)C(C)(C)O2)C2=CC=CC=C12 | |
430.16 | |
430.16 | |
Anthracene-9,10-diboronic acid bis(pinacol) ester |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.