missing translation for 'onlineSavingsMsg'
Learn More

Anthracene-9-carboxylic acid, 98+%

Product Code. 11447956
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11447956

Brand: Thermo Scientific Alfa Aesar A14049.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 723-62-6 | C15H10O2 | 222.24 g/mol

Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.

Solubility
Soluble to 10 mm in ethanol and to 100 mm in DMSO. Soluble in benzene, chloroform, and hydronaphthalenes. Insoluble in water.

Notes
Store in room temperature. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 723-62-6
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.24
MDL Number MFCD00001257
InChI Key XGWFJBFNAQHLEF-UHFFFAOYSA-N
Synonym 9-anthracenecarboxylic acid, 9-anthroic acid, anca, 9-carboxyanthracene, 9-ac, anthracene-10-carboxylic acid, 9-anthracene carboxylic acid, a9c, spectrum_001457, tocris-0963
PubChem CID 2201
ChEBI CHEBI:34507
IUPAC Name anthracene-9-carboxylic acid
SMILES OC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point ∼218°C (decomposition)
Quantity 10 g
Beilstein 1875336
Solubility Information Soluble to 10 mm in ethanol and to 100 mm in DMSO. Soluble in benzene,chloroform,and hydronaphthalenes. Insoluble in water.
Formula Weight 222.24
Percent Purity ≥98%
Chemical Name or Material Anthracene-9-carboxylic acid
compliance-icons
Product Identifier
  • Anthracene-9-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 9-Anthracenecarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.