Learn More
Anthrone, ACS, Thermo Scientific Chemicals
Used for determination of carbohydrates
£50.30 - £142.00
Chemical Identifiers
CAS | 90-44-8 |
---|---|
Molecular Formula | C14H10O |
Molecular Weight (g/mol) | 194.23 |
MDL Number | MFCD00001187 |
InChI Key | RJGDLRCDCYRQOQ-UHFFFAOYSA-N |
Synonym | anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744 |
PubChem CID | 7018 |
ChEBI | CHEBI:33835 |
IUPAC Name | 10H-anthracen-9-one |
SMILES | O=C1C2=CC=CC=C2CC2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11367338
|
Thermo Scientific Alfa Aesar
030739.14 |
25 g |
£50.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357338
|
Thermo Scientific Alfa Aesar
030739.22 |
100 g |
£142.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used for determination of carbohydrates Anthrone is used for the determination of carbohydrates. It is used as a general reagent in the colorimetric determination of carbohydrates in yeast and plant extracts. It is involved in organic synthesis. It is utilized for the determination of sugar in body fluids. It finds application as a laxative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
90-44-8 | |
194.23 | |
RJGDLRCDCYRQOQ-UHFFFAOYSA-N | |
7018 | |
10H-anthracen-9-one |
C14H10O | |
MFCD00001187 | |
anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744 | |
CHEBI:33835 | |
O=C1C2=CC=CC=C2CC2=CC=CC=C12 |
Specifications
90-44-8 | |
C14H10O | |
1910173 | |
14,691 | |
Soluble in acetone,hot benzene,concentrated sulfuric acid,hot dilute alkali and most organic solvents. Insoluble in water. | |
O=C1C2=CC=CC=C2CC2=CC=CC=C12 | |
194.23 | |
CHEBI:33835 | |
ACS | |
Anthrone |
155°C to 158°C | |
MFCD00001187 | |
Light sensitive | |
anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744 | |
RJGDLRCDCYRQOQ-UHFFFAOYSA-N | |
10H-anthracen-9-one | |
7018 | |
194.23 | |
Crystalline |
Safety and Handling
GHS H Statement
H315-H319-H335
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 201-994-0
missing translation for 'rtecsNumber' : CB8925500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only