missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-AKT1, CHUK Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0097.1Set
Includes: AKT1 rabbit purified polyclonal antibody (20μg); CHUK mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Descripción
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the AKT1 protein, and the other against the CHUK protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Especificaciones
| AKT1 | |
| Monoclonal/Polyclonal | |
| AKT1 | |
| AKT1 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. AKT1 rabbit purified polyclonal antibody (20 ug) 2. CHUK mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
| Purified |
| In situ PLA | |
| Unconjugated | |
| AKT/MGC99656/PKB/PKB-ALPHA/PRKBA/RAC/RAC-ALPHA | |
| Mouse, Rabbit | |
| RUO | |
| 207/1147 | |
| Human | |
| AKT1 & CHUK Protein Protein Interaction Antibody Pair |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido