missing translation for 'onlineSavingsMsg'
Learn More

ALDH4A1, Human, Matched Antibody Pair (AP51), Abnova™

Brand:  Abnova H00008659-AP51.1Set

Additional Details : Weight : 0.00010kg

Product Code. 16040789

  • £660.00 / 1 set
Estimated Shipment: 21-08-2024
to see stock.

Includes: Capture antibody: mouse monoclonal anti-ALDH4A1 (100μg); Detection antibody: rabbit purified polyclonal anti-ALDH4A1 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.

Description

Description

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ALDH4A1.

Host Species: Mouse, Rabbit
Species Reactivity: Hu

ELISA Pair (Transfected Lysate)

Specifications

Specifications

ALDH4A1
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ALDH4A1.
ALDH4/P5CD/P5CDh/P5CDhL/P5CDhS
Mouse, Rabbit
RUO
8659
Human
Purified
ELISA
Unconjugated
ALDH4A1
ALDH4A1
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-ALDH4A1 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-ALDH4A1 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers