missing translation for 'onlineSavingsMsg'
Learn More

alkB, alkylation repair homolog 1 (E. coli), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ALKBH protein.

Brand:  Abnova H00008846-B02.50uL

Product Code. 16105358

  • £283.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This gene encodes a homolog to the E. coli alkB gene product. The E. coli alkB protein is part of the adaptive response mechanism of DNA alkylation damage repair. It is involved in damage reversal by oxidative demethylation of 1-methyladenine and 3-methylcytosine. [provided by RefSeq

Sequence: MGKMAAAVGSVATLATEPGEDAFRKLFRFYRQSRPGTADLEGVIDFSAAHAARGKGPGAQKVIKSQLNVSSVSEQNAYRAGLQPVSKWQAYGLKGYPGFIFIPNPFLPGYQWHWVKQCLKLYSQKPNVCNLDKHMSKEETQDLWEQSKEFLRYKEATKRRPRSLLEKLRWVTVGYHYNWDSKKYSADHYTPFPSDLGFLSEQVAAACGFEDFRAEAGILNYYRLDSTLGIHVDRSELDHSKPLLSFSFGQSAIFLLGGLQRDEAPTAMFMHSGDIMIMSGFSRLLNHAVPRVLPNPEGEGLPHCLEAPLPAVLPRDSMVEPCSMEDWQVCASYLKTARVNMTVRQVLATDQNFPLEPIEDEKRDISTEGFCHLDDQNSEVKRARINPHS
Specifications

Specifications

alkB, alkylation repair homolog 1 (E. coli)
Polyclonal
Mouse polyclonal antibody raised against a full-length human ALKBH protein.
ALKBH1
ABH/ABH1/ALKBH/alkB/hABH
Mouse
50 μL
Yes
8846
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Western Blot
Unconjugated
No additive
BC025787.1
ALKBH1
ALKBH (AAH25787.1, 1 a.a. ∼ 389 a.a) full-length human protein.
RUO
Primary
Human
Serum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.