Learn More
anti-ART3 Matched Antibody Pair, Abnova
Brand: Abnova H00000419-AP11.1Set
Additional Details : Weight : 0.00010kg
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-ART3 (100μg); Mouse monoclonal anti-ART3, IgG2bκ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
ADP-ribosylation is a reversible posttranslational modification used to regulate protein function. ADP-ribosyltransferases, such as ART3, transfer ADP-ribose from NAD+ to the target protein, and ADP-ribosylhydrolases.
ELISA
Specifications
ART3 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ART3. | |
FLJ26404 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-ART3 (100 μg) 2. Detection antibody: mouse monoclonal anti-ART3, IgG2b Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
ART3 | |
ART3 | |
Affinity Purified | |
RUO | |
419 | |
Human | |
IgG2b |