missing translation for 'onlineSavingsMsg'
Learn More

ASNA1, Mouse, Clone: 2H3, Abnova™

Mouse monoclonal antibody raised against a partial recombinant ASNA1.

Brand:  Abnova H00000439-M03.100ug

Product Code. 16184944

  • £283.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ASNA1 is the human homolog of the bacterial arsA gene. In E. coli, ArsA ATPase is the catalytic component of a multisubunit oxyanion pump that is responsible for resistance to arsenicals and antimonials.[supplied by OMIM

Sequence: PEQTTFICVCIAEFLSLYETERLIQELAKCKIDTHNIIVNQLVFPDPEKPCKMCEARHKIQAKYLDQMEDLYEDFHIVKLPLLPHEVRGADKVNTFSALLLEPYKPPSAQ
Specifications

Specifications

ASNA1
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_004317
ASNA1
ASNA1 (NP_004308, 239 a.a. ∼ 348 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG1 κ
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), KnockDown, Western Blot
2H3
Mouse monoclonal antibody raised against a partial recombinant ASNA1.
ASNA1
ARSA-I/ARSA1/MGC3821
Mouse
Affinity chromatography
RUO
439
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.