missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-ATF4, JUN Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0607.1Set
Additional Details : Weight : 0.00010kg
Includes: ATF4 rabbit purified polyclonal antibody (20μg); JUN mouse purified polyclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
Host Species: Rabbit, Mouse
Cross Reactivity: Human
In situ PLA
Specifications
ATF4 | |
Monoclonal/Polyclonal | |
ATF4 | |
ATF4 | |
1 Set | |
468/3725 | |
Human | |
ATF4 & JUN Protein Protein Interaction Antibody Pair |
In situ PLA | |
Unconjugated | |
CREB-2/CREB2/TAXREB67/TXREB | |
Mouse, Rabbit | |
Primary | |
Antibody pair set content: 1. ATF4 rabbit purified polyclonal antibody (20 ug) 2. JUN mouse purified polyclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |