missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Anti-bacterial, Anti-odor Insoles
Double-action, perforated blue insoles prevent bacteria and odor.
£66.56 - £68.76
Specifications
Color | Blue |
---|---|
For Use With (Application) | Insoles for comfort, prevent bacteria/odor |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15342790
|
Honeywell™
6000310-38/7 |
T38 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15342800
|
Honeywell™
6000310-43/7 |
T43 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15352800
|
Honeywell™
6000310-44/7 |
T44 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Dual-action: prevents bacteria and odor
- Perforated for breathability
Specifications
Blue | |
Insoles for comfort, prevent bacteria/odor |