missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Anti-bacterial, Anti-odor Insoles

Product Code. 17415043
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 17415043

Brand: Honeywell™ 600031047/7

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Double-action, perforated blue insoles prevent bacteria and odor.

  • Dual-action: prevents bacteria and odor
  • Perforated for breathability
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Design Perforated
For Use With (Application) Preventing Bacteria and Odor
Size 47
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.