missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Anti-bacterial, Anti-odor Insoles
Double-action, perforated blue insoles prevent bacteria and odor.
£10.60 - £10.85
Specifications
Color | Blue |
---|---|
Design | Perforated |
For Use With (Application) | Preventing Bacteria and Odor |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17425033
|
Honeywell™
6000310-38/7 |
38 |
£10.60
1 pair |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
17435033
|
Honeywell™
6000310-39/7 |
39 |
£10.60
1 pair |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
17445033
|
Honeywell™
6000310-40/7 |
40 |
£10.60
1 pair |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
17455033
|
Honeywell™
6000310-41/7 |
41 |
£10.60
1 pair |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
17465033
|
Honeywell™
6000310-42/7 |
42 |
£10.85
1 pair |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
17475033
|
Honeywell™
6000310-43/7 |
43 |
£10.85
1 pair |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
17485033
|
Honeywell™
6000310-44/7 |
44 |
£10.60
1 pair |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
17495033
|
Honeywell™
6000310-45/7 |
45 |
£10.60
1 pair |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
17405043
|
Honeywell™
6000310-46/7 |
46 |
£10.60
1 pair |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
17415043
|
Honeywell™
6000310-47/7 |
47 |
£10.85
1 pair |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
Description
- Dual-action: prevents bacteria and odor
- Perforated for breathability
Specifications
Blue | |
Perforated | |
Preventing Bacteria and Odor |